Learn More
DWK Life Sciences Wheaton™ Crimpenstein™ Electric Crimper/Decapper - Non-Standard Voltage
Shop All DWK Life Sciences Products
Description
Includes
Each Crimpenstein comes with a detachable power cord, a convenient carrying/storage case, a crimper and deapper jaw set or an individual crimper or decapper jaw.
Wheaton Crimpenstein Electric Crimper/Decapper
- Easily attaches and removes aluminum seals on crimp top/serum finish bottles and vials to improve productivity
- Adjustable jaw intensity with memory allows user to regulate crimper and decapper jaw action
- Quick-change jaw-sets available in 11, 13 and 20mm sizes allow for crimping and decapping of different size bottles and vials
- Lightweight and easy to operate enables semiautomatic operation for extended periods with less effort
- Electric powered for quick setup and high volume operation
Optional sturdy stand (Part No. W225630) holds the Crimpenstein in a fixed vertical position for operations where vials need to be handled one at a time.
Order Info
Crimper and Decapper Jaws (Part No. W225616-W225628) and sturdy stand (Part No. W225630) are sold separately. For standard voltages, see cat. no. 029110001.
Specifications
Specifications
| For Use With (Application) | Attaching and removing aluminum seals on crimp top/serum finish bottles and vials |
| Includes | Italy, Chile Plug |
| For Use With (Equipment) | 13 mm Flip-cap Aluminum Seals |
| Product Type | 13 mm Crimper/Decapper Set |