Learn More
DWK Life Sciences™ Wheaton™ Crimpenstein™ Electric Crimper/Decapper - Non-Standard Voltage
Lightweight and easy to operate
Manufacturer:  DWK Life Sciences™ W225638D
 View more versions of this product
Catalog No. 02-912-833
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Includes: Each Crimpenstein comes with a detachable power cord, a convenient carrying/storage case, a crimper and deapper jaw set or an individual crimper or decapper jaw.
Description
Description
Wheaton Crimpenstein Electric Crimper/Decapper
  • Easily attaches and removes aluminum seals on crimp top/serum finish bottles and vials to improve productivity
  • Adjustable jaw intensity with memory allows user to regulate crimper and decapper jaw action
  • Quick-change jaw-sets available in 11, 13 and 20mm sizes allow for crimping and decapping of different size bottles and vials
  • Lightweight and easy to operate enables semiautomatic operation for extended periods with less effort
  • Electric powered for quick setup and high volume operation
For information on using the Crimpenstein with flip cap aluminum seals, visit www.wheatonsci.com/crimp. For information on Wheaton crimp top/serum finish bottles and vials, aluminum seals and rubber stoppers, visit www.fishersci.com.
Optional sturdy stand (Part No. W225630) holds the Crimpenstein in a fixed vertical position for operations where vials need to be handled one at a time.
Order Info
Crimper and Decapper Jaws (Part No. W225616-W225628) and sturdy stand (Part No. W225630) are sold separately. For standard voltages, see cat. no. 029110001.
Specifications
Specifications
Attaching and removing aluminum seals on crimp top/serum finish bottles and vials
20mm Flip-cap Aluminum Seals
Crimpenstein with 20mm Crimper Jaw; U.K. plug