Learn More
DWK Life Sciences Wheaton™ Cryule™ Cryogenic Ampules
For biological sample preservation in glass.
$359.47
Specifications
Product Type Unscored Cryogenic Ampule
Certifications/Compliance USP Type I, ASTM E438 Type I, Class A
View More Specs
Products 1
Catalog Number Mfr. No. Capacity (Metric) Height (Metric) Diameter (Metric) Outer Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (Metric) Height (Metric) Diameter (Metric) Outer Price Quantity & Availability  
13-709-131
View Documents Promotion Details
DWK Life Sciences
W651469
5mL 98mm 16.5cm
Case of 293 for $359.47
Add to cart
Description
Description
  • Allows for storage at low temperatures
  • Made from low extractable borosilicate glass that conforms to USP Type I and ASTM E 438 Type I, Class A requirements
  • Can be used in the vapor phase of liquid nitrogen
  • Special design allows for storage at low temperatures as well as rapid thawing
Certifications
USP Type I, ASTM E438 Type I, Class A
Specifications
Specifications
Unscored Cryogenic Ampule
USP Type I, ASTM E438 Type I, Class A