Learn More
Wheaton™ Cryule™ Gold-Band™ Cryogenic Ampules Available on GSA/VA Contract for Federal Government customers only.
For preservation of biological materials with liquid nitrogen
Manufacturer:  Wheaton™ 651483
 View more versions of this product
Catalog No. 06-401C
  Check Availability
Add to cart
Alert:
Cryogenic vials are intended for placement only in the vapor phase of liquid nitrogen, and should not be used for storage in the liquid phase of liquid nitrogen. Immersion of the vials in the liquid phase could result in penetration of the liquefied gas into the vial, resulting in rapid vaporization of the liquid upon removal and possible violent explosion or leakage from the vial/closure perimeter.
To prevent cryogenic vials from exploding, never overfill liquid nitrogen storage units. Always examine vials before use to ensure no visible defects around the closure rims. Always use full faceshields, heavy safety gloves and laboratory protective apparel when removing vials from cryogenic storage. Always permit vials to warm slowly in a biological safety cabinet. Never reuse cryogenic vials.
Specifications Description & Specifications
Specifications
1.2mL
Clear
0.19 in.
0.5cm
5.8cm
Prescored Cryogenic Ampule
ASTM E438 Type I
0.46 in.
1.18cm
2.28 in.
Borosilicate Glass
Accessories