Learn More
DWK Life Sciences Wheaton™ Culture Vessel
Perform repeated wet or dry sterilization to these culture vessels, which are made of borosilicate glass
Manufacturer:  DWK Life Sciences 348528
 View more versions of this product
Catalog No. 02-913-303
Add to cart
Description
Description
  • Conforms to USP and ASTM requirements
  • 38, 45 or 51mm screw head
  • 2.6mm wall thickness
  • Bottle material made of borosilicate glass
  • Autoclavable
  • Reusable
Specifications
Specifications
110mm
535mm