Wheaton™ Disposable 1000µL Pipette Microtips Available on GSA/VA Contract for Federal Government customers only.
Premium pipette tips covering most research and routine applications, compatible with most pipettes
Manufacturer: Wheaton™ 851276
 View more versions of this product
Catalog No. 13-707-71
  Check Availability
Add to Cart
    Specifications
    Product Type Micro Tip
    Material Virgin Polypropylene
    Sterility Nonsterile
    Tip Style Disposable
    Packaging Bulk pack of 1000 tips
    Quantity 1000 Case
    Certifications/Compliance RNase, DNase-free
    For Use With (Application) Premium pipette tips covering most research and routine applications, compatible with most pipettes, excellent accuracy and reproducibility
    Volume (Metric) 1 to 1000μL
    Color Blue
  • Description & Specifications
Related Products
Product Content Feedback