Wheaton™ Disposable 200µL Pipette Microtips Available on GSA/VA Contract for Federal Government customers only.

Premium pipette tips covering most research and routine applications, compatible with most pipettes

Manufacturer: Wheaton™ 851271

 View more versions of this product

Catalog No. 13-707-69

  Check Availability
Add to Cart

    Specifications

    Product Type Micro Tip
    Material Polypropylene
    For Use With (Equipment) Wheaton Calibra™ and Acura™ fixed and adjustable-volume pipets
    Sterility Nonsterile
    Autoclavable Yes
    Tip Style Disposable
    Packaging Bulk
    Certifications/Compliance DNase, RNase, pyrogen free
    Volume (Metric) 200μL
    Color Yellow

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit