Learn More
Wheaton™ Disposable Pasteur Pipets
Manufacturer:  Wheaton™ 357335
 View more versions of this product
Catalog No. 13-678-8D
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Specifications Description & Specifications
Specifications
USP type I, ASTM E 438 type I, Class A
0.25 to 0.29 in.
Yes
Constriction near the mouth end
4.25 in.
12cm
22.8cm
For inoculating culture media and as a general sampling pipet, can also used in applying samples in chromatography
Pasteur Pipet
Clear
0.65 to 0.75cm
Acura™ manual 835 macro pipettes or Profiller™ electro 446
4.72 in.
8.97 in.
10.8cm
Straight
Borosilicate Glass