Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ DualFusion Vials

DWK Life Sciences Wheaton™ DualFusion vials protect samples from container leachates and environmental stressors.

$214.00 - $400.00

Specifications

Autoclavable Yes
Certifications/Compliance Certificate of Analysis
Color Clear
Disposable Yes
For Use With Snap Caps or Rubber Stoppers and Aluminum Seals
View More Specs

Products 3
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
Catalog Number Mfr. No. Volume (Metric) Price Quantity & Availability  
15-112-129 View Documents DWK Life Sciences
W986000DFS
2 mL Case of 100 for $400.00
Add to cart
 
15-112-130 View Documents DWK Life Sciences
W986001DFS
6 mL Case of 48 for $214.00
Add to cart
 
15-112-131 View Documents DWK Life Sciences
W986002DFS
10 mL Case of 48 for $214.00
Add to cart
 
Description

Description

  • Wheaton™ DualFusion vials combine the best properties of glass, with the mechanical strength and the precision molding of plastic.
  • Manufactured in a Class 7 Clean Room and E-Beam sterilized to meet USP788 for low particulate levels and USP85 for low endotoxin level for injectables
  • Engineered to protect pharmaceutical drugs and sample formulations that are prone to cause glass delamination and that are sensitive to oxidation via the permeation of atmospheric oxygen.
  • Using a state-of-the-art Plasma Enhanced Chemical Vapor Deposition (PECVD) Technology, an organosilicate protective layer is fused with a silica-like (SiO2 barrier) layer.
  • These layers are fused to a Cyclic Olefin Polymer (COP) shell to form a robust covalently bonded material resulting in an inert surface.
  • DualFusion stand-alone nested tubs can be paired with Snap Cap and Aluminum Seal closure offerings
  • Wheaton™ DualFusion Vials are manufactured by and sold under license from SiO2 Medical Products, Inc.
Specifications

Specifications

Yes
Clear
Snap Caps or Rubber Stoppers and Aluminum Seals
Vials Only
COP SI02 DualFusion
Sterile
Certificate of Analysis
Yes
No
No
SiO2
Serum Vial
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit