Learn More
Wheaton™ E-C Borosilicate Glass Screw Thread Sample Vials with Rubber-Lined Caps Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Wheaton™ 224720
 View more versions of this product
Catalog No. 03-343-3A
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Yes
Screw cap
12mm
Clear Low Extractable Borosilicate Glass
1.8mL
35mm
14B Rubber
Screw
USP Type I, ASTM E438, Type I, Class A
1.5mL
Clear
E-C Sample Vial
8-425
Phenolic
GPI 8-425
0.5 dr.