Learn More
Wheaton™ E-C Borosilicate Glass Screw Thread Sample Vials with Rubber-Lined Caps Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  Wheaton™ 224722
 View more versions of this product
Catalog No. 03-343-3C
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Yes
Screw cap
15mm
Clear Low Extractable Borosilicate Glass
4mL
48mm
14B Rubber
Screw
USP Type I, ASTM E438, Type I, Class A
4.0mL
Clear
E-C Sample Vial
13-425
Phenolic
GPI 13-425
1.08 dr.