Learn More
DWK Life Sciences Wheaton™ E-C Borosilicate Glass Screw Thread Sample Vials with Rubber-Lined Caps Available on GSA/VA Contract for Federal Government customers only.
Manufacturer:  DWK Life Sciences 224724
 View more versions of this product
Catalog No. 03-343-3E
  Check Availability
Add to cart
Description
Description
  • Clear vials are manufactured from Wheaton 33 low extractable borosilicate glass that conforms to ASTM Type I, Class A and USP Type I requirements
  • Amber vials are manufactured from Wheaton 320 amber glass that conforms to USP Type I requirements to protect light-sensitive samples
  • Vials and screw caps packaged separately
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
Yes
Screw cap
17mm
Clear Low Extractable Borosilicate Glass
8mL
63mm
14B Rubber
Screw
USP Type I, ASTM E438, Type I, Class A
8.0mL
Clear
E-C Sample Vial
15-425
Phenolic
GPI 15-425
2.2 dr.