Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ E-C Borosilicate Glass Screw Thread Sample Vials without Caps

Manufactured from Wheaton 33 low extractable borosilicate glass. Wheaton™ E-C Borosilicate Glass Screw Thread Sample Vials without Caps are economically priced and conveniently packed in two shrink-wrapped packs which reduces contamination from corrugated material found in traditional packaging.

Manufacturer:  DWK Life Sciences 224711

 View more versions of this product

Catalog No. 02-985-132


  Check Availability
Add to cart

Description

Description

  • Caps are packed separately
  • Choice of amber or clear glass
Specifications

Specifications

Yes
Screw cap
0.5 dr., 2mL
Amber
Borosilicate Glass
2mL
Screw
USP Type I
2mL
12mm
0.5 dr.
E-C Sample Vial
35mm
0.5 dr.
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit