Learn More
Wheaton™ E-C Borosilicate Glass Screw Thread Sample Vials without Caps Available on GSA/VA Contract for Federal Government customers only.
Manufactured from Wheaton 33 low extractable borosilicate glass. Wheaton™ E-C Borosilicate Glass Screw Thread Sample Vials without Caps are economically priced and conveniently packed in two shrink-wrapped packs which reduces contamination from corrugated material found in traditional packaging.
Manufacturer:  Wheaton™ 224706
 View more versions of this product
Catalog No. 03-343G
  Check Availability
Add to cart
Specifications
Description
  • Caps are packed separately
  • Choice of amber or clear glass
Description & Specifications
Specifications
Yes
Screw cap
4 dr., 16.0mL
Clear
Low Extractable Borosilicate Glass
16mL
Screw
USP Type I, ASTM E438, Type I, Class A
16.0mL
21mm
4 dr.
E-C Sample Vial
70mm
4.3 dr.