Learn More
Wheaton™ E-Z Decappers Available on GSA/VA Contract for Federal Government customers only.
With ergonomically designed handles for sealing and decapping standard laboratory vials
Manufacturer:  Wheaton™ W225352
 View more versions of this product
Catalog No. 06-451-462
  Check Availability
Add to cart
Specifications
Description
Wheaton E-Z Decappers are recommended for decapping standard laboratory vials.
  • Several sizes of decapper are available, including those that remove seals 8mm, 11mm, 13mm and 20mm wide.
  • The decappers come with visible size labels for easy identification.
  • User comfort is preserved by an ergonomically designed, cushioned handle. Hand fatigue is reduced by the short handle and redesigned spring assembly.
  • Decappers can be autoclaved, although autoclaving may change the appearance or reduce product life. They should not be autoclaved at greater than 121°C
Description & Specifications
Specifications
13mm Seals
Seal Decapper