Wheaton™ E-Z Seals™ Aluminum Vial Caps Available on GSA/VA Contract for Federal Government customers only.
Manufacturer: Wheaton™ 22421901
 View more versions of this product
Catalog No. 06-406-24
  Check Availability
Add to Cart
    Specifications
    Closure Type Open Top
    Color Natural
    Quantity 100 Pack
    Material Closure Aluminum
    Thread Size 11mm
    Closure Material Aluminum
    Type E-Z Seals™
    Material (Liner) PTFE/Silicone
  • Description & Specifications
Related Products
Product Content Feedback