Learn More
DWK Life Sciences Wheaton™ Glass Diagnostic Vials with Screw Threads
Alternative to serum bottles and vials
Manufacturer:  DWK Life Sciences W216376
 View more versions of this product
Catalog No. 02-912-453
Add to cart
Description
Description
Ideal for lyophilization.
  • Screw-thread design eliminates the need for crimping
  • Fits with I-Loc™ closure or screw cap with thin flange stopper
  • Tubular design provides excellent clarity and dimensional consistency from vial to vial
  • Heavy-walled glass provides impact resistance
Specifications
Specifications
Type 1
Diagnostic Vials
Amber
Screw Thread
0.34
10mL