Learn More
DWK Life Sciences Wheaton™ Glass Sample Vials in Lab File without Caps
SDP
Packaged in partitioned trays, without caps
Supplier:  DWK Life Sciences 224802
 View more versions of this product
Catalog No. 06-412-2
Add to Cart
Description
Description
Partitioned trays provide an easy way to inventory samples or to store empty vials.
  • Autoclavable
  • Clear or Amber Vials
    • Clear vials made from low-extractable borosilicate glass that provides superior chemical resistance
    • Amber vials made from borosilicate glass that protects light-sensitive products
    Order Info
    Caps available separately
    Compliance
    Clear vials conform to USP Type 1 and ASTM™ E 438 Type 1, Class A requirements. Amber vials conform to USP Type 1 requirements for light transmission.
    Specifications
    Specifications
    Clear
    Borosilicate Glass
    Screw
    Autoclavable
    45 mm
    M-T File™ and Vial Rack
    15 mm
    4 mL
    1.08 dr.
    Screw Cap
    ASTM E438, USP Type I, Type I, Class A
    Product Suggestions
    Product Suggestions