Learn More
DWK Life Sciences Wheaton™ Glass Sample Vials in Lab File without Caps
Packaged in partitioned trays, without caps
Manufacturer:  DWK Life Sciences 224811
 View more versions of this product
Catalog No. 06-412-7
Add to cart
Description
Description
Partitioned trays provide an easy way to inventory samples or to store empty vials.
  • Autoclavable
  • Clear or Amber Vials
    • Clear vials made from low-extractable borosilicate glass that provides superior chemical resistance
    • Amber vials made from borosilicate glass that protects light-sensitive products
    Specifications
    Specifications
    Screw
    USP Type I
    12 mm
    Standard Glass Vial
    M-T File™ and vial rack
    Screw
    Yes
    Amber
    Borosilicate Glass
    2 mL
    35mm
    0.5 dr.