Learn More
DWK Life Sciences Wheaton™ Glass Sample Vials in Vial File™ Available on GSA/VA Contract for Federal Government customers only.
Vial File provides a compact, easily accessible means of sample storage
Manufacturer:  DWK Life Sciences 224950
 View more versions of this product
Catalog No. 03-336-6
Add to cart
Description
Description
Caps attached to vials.
  • Polystyrene case with partitions, foam inserts and alphanumeric indexing card helps organize and protect vials
  • Choice of solid- or open-top black phenolic caps: open caps facilitate sample retrieval with syringe
  • Vials and caps autoclavable
Clear or Amber Vials
  • Clear vials made from low-extractable borosilicate glass that provides superior chemical resistance
  • Amber vials made from borosilicate glass that protects light-sensitive products
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Compliance
Clear vials conform to USP Type 1 and ASTM™ E 438 Type 1, Class A requirements. Amber vials conform to USP Type 1 requirements of light transmission.
Specifications
Specifications
Yes
Phenolic
Used for storing and transporting vials
38mm
Screw
0.405 dr.
12mm
1.5mL
USP Type I, ASTM E438, Type I, Class A
Open top screw cap
No
PTFE/Silicone
Glass Sample Vial
Clear
Borosilicate Glass (Low Extractable)