Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Headspace Vials Convenience Kits with Pressure Release Available on GSA/VA Contract for Federal Government customers only.

Newly designed top provides a tighter seal with PTFE-silicone septa

Manufacturer:  DWK Life Sciences W22527801

 View more versions of this product

Catalog No. 02-911-313


  Check Availability
Add to cart

Description

Description

  • Manufactured from Type I borosilicate glass
  • Rounded shoulders and bottoms allow for even heating and safer operation at higher pressures

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Order Info

Available in convenience packs of 100. Dia. x H (10mL vials): 23x 46mm. Dia. x H (20mL vials): 23 x 75mm.

Specifications

Specifications

Autosampler Vial
10mL
Clear
46mm
23mm
6.0mL
20mm
Borosilicate Glass
Crimp
100 Case
1.81 in.
Documents
Accessories
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit