Learn More
Wheaton™ HPLC Mobile-Phase Reservoirs Available on GSA/VA Contract for Federal Government customers only.
Suitable for use as mobile phase and filtration reservoirs, as well as general sample and buffer storage
Manufacturer:  Wheaton™ 264710
 View more versions of this product
Catalog No. 13-703-993
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Flat
2000mL
USP Type I and ASTM E 438 Type I, Class A
138mm
Ideal for mobile phase or filtration reservoir
246mm
HPLC Mobile-phase Reservoir
67.62 oz.
67.62 oz., 2000mL
5.43 in.
Screw thread
9.68 in.
Glass