Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Hybridization Bottles
SDP

Catalog No. p-42660
Click to view available options
Length (Metric):
75 mm
100 mm
150 mm
300 mm
Diameter (Metric) Inner:
35 mm
70 mm
Product Type:
Hybridization Bottle
Safety-coated Hybridization Bottle

For use in nucleic acid and protein blotting procedures that require a minimum of probe solution

  • Manufactured from heavy-wall borosilicate glass
  • Safety-coated bottles with strong, slip-resistant plastisol covers reduce potential breakage and exposure to hazardous materials
  • Polypropylene caps incorporate sonically bonded PTFE-fluorocarbon resin/silicone lining to prevent leakage
  • Acrylic rack holds up to six hybridization bottles of different sizes for safe transport or storage

Order Info

Items come as one per case

TRUSTED_SUSTAINABILITY

Specifications

Closure Type Screw Cap
Product Type Hybridization Bottle
Closure Size 45 mm
Length (Metric) 150 mm
Diameter (English) Inner 1.37 in.
Diameter (Metric) Inner 35 mm
Material Borosilicate Glass
Length (English) 5.90 in.
Coating Plastisol
Liner Material PTFE or Silicone
Certifications/Compliance USP Type I
For Use With (Application) Nucleic acid and protein blotting procedures that require a minimum of probe solution
Closure Material Polycarbonate
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.