Learn More
Wheaton™ Hybridization Bottles and Racks Available on GSA/VA Contract for Federal Government customers only.
For use in nucleic acid and protein blotting procedures that require a minimum of probe solution
Manufacturer:  Wheaton™ 805007
 View more versions of this product
Catalog No. 13-245-97D
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
USP Type I
45mm
1.37 in.
Nucleic acid and protein blotting procedures that require a minimum of probe solution
PTFE or Silicone
Screw
Hybridization Bottle
Polycarbonate
Plastisol
35mm
11.81 in.
Borosilicate Glass
300mm