Learn More
Wheaton™ I-LOC Closures Available on GSA/VA Contract for Federal Government customers only.
Eliminate use of separate aluminum seal and rubber stopper
Manufacturer:  Wheaton™ 24067603
 View more versions of this product
Catalog No. 06-440-50
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Screw
Red
I-Loc™ Cap
Bromobutyl
Yes
Polypropylene
Polypropylene
20-400