Learn More
DWK Life Sciences Wheaton™ Impellers for Celstir™ Spinner Flasks
$174.00 - $200.00
Specifications
Includes Top cap with liners, glass rod, magnet holder, magnet and stirring paddle
View More Specs
Products 4
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
Catalog Number Mfr. No. For Use With (Equipment) Price Quantity & Availability  
06-416-5C View Documents DWK Life Sciences
356896
125mL Celstir™ Spinner Flask Case for $174.00
Add to cart
06-416-5D View Documents DWK Life Sciences
356899
250mL Celstir™ Spinner Flask Case for $174.00
Add to cart
06-416-5E View Documents DWK Life Sciences
356902
500mL Celstir™ Spinner Flask Case for $197.00
Add to cart
06-416-5F View Documents DWK Life Sciences
356904
1000mL Celstir™ Spinner Flask Case for $200.00
Add to cart
Description
Description
  • This non-breakable stainless steel shaft, for replacing the glass shaft included with Celstir™ flasks, may be retrofitted to the existing Celstir flasks.
Specifications
Specifications
Top cap with liners, glass rod, magnet holder, magnet and stirring paddle