Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wheaton™ LDPE Dropping Bottles Available on GSA/VA Contract for Federal Government customers only.

Excellent chemical resistance

Manufacturer:  Wheaton™ W242824A

 View more versions of this product

Catalog No. 03-439-76


  Check Availability
Add to cart

Specifications

Description

  • Available in a variety of sizes
  • Packaged in cases of 100, 1000 or bulk

Description & Specifications

Specifications

10mL, 0.34 oz.
0.34 oz.
Natural
1.77 in.
1000/Cs.
No
15-415
45mm
10mL
Cellulose Acetate Proprionate
1 in.
LDPE
1000/Cs.
No
25mm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit