Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wheaton™ LDPE Dropping Bottles Available on GSA/VA Contract for Federal Government customers only.

Excellent chemical resistance

Manufacturer:  Wheaton™ W242831

 View more versions of this product

Catalog No. 03-439-71H


  Check Availability
Add to cart

Specifications

Description

  • White LDPE helps protect UV- lght-sensitivie samples
  • Packaged in cases of 100, 1000 or bulk

Description & Specifications

Specifications

3mL, 0.1 oz.
0.1 oz.
0.51 in.
LDPE
100/Cs.
No
14mm
3mL
White
1.54 in.
100/Cs.
No
8-425
39mm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit