Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wheaton™ LDPE Dropping Bottles with Extended Controlled Dropper Tips and Polypropylene Dropper Tip Caps Available on GSA/VA Contract for Federal Government customers only.

Extended controlled dropper tip dispenses one drop at a time

Manufacturer:  Wheaton™ 211623

 View more versions of this product

Catalog No. 03-439-61Q


  Check Availability
Add to cart

Specifications

Description

Average drop size: 44µL ±6µL

  • Radiation-grade polypropylene screw caps and tips packaged separately
  • Available in natural or white tips, packaged separately

Description & Specifications

Specifications

0.51 oz., 15mL
0.51 oz.
White
Extended controlled dropper tip
LDPE
No
25mm
15mL
Polypropylene
1 in.
3.07 in.
Yes
15-415
78mm
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit