Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Low-Density Extended Controlled Dropper Tips (13mm)

Click to view available options
Color:
Blue
Gray
Green
Natural
Orange
Pink
Purple
Red
White
Yellow
Material:
LDPE
Polyethylene
No. per Case:
100 Cs.
1000 Cs.
  • Extended controlled dropper tip dispenses one drop at a time
  • Average drop size is 44μL ±6μL
  • Case of 100
Certifications/Compliance ISO 9001
Color Purple
Drop Size 41μL
Material LDPE
No. per Case 100 Cs.
Type Dropper Tips
For Use With (Equipment) Plastic dropping bottle