Learn More
DWK Life Sciences Wheaton™ Micro Tissue Grinders
Used in extremely precise work when delicate hand operation is required
Manufacturer:  DWK Life Sciences 357848
 View more versions of this product
Catalog No. 08-414-15B
Add to cart
Description
Description
  • Made from low extractable borosilicate glass
Potter-Elvehjem Micro Tissue Grinder
  • Reservoir and pouring lip provide easy decanting of micro sample
  • Unit can be autoclaved / sterilized to prevent cross contamination between samples
  • Pestle clearance range: 0.1 – 0.15mm
Micro Tissue Grinder with PTFE Spacer
  • PTFE spacer aligns the pestle and reduces aerosoling of the product
  • Includes a finely ground glass pestle and tube
  • Autoclavable
Disposable Micro Tissue Grinder
  • Disposable to eliminate cross contamination and useful for radioactive samples
  • Small grinder area allows for high sample recovery
Specifications
Specifications
Yes
0.2mL
Micro disposable tissue grinder
4.52 in.
1.2 in.
0.27 in.
0.006 oz., 0.2mL
0.006 oz.
Borosilicate Glass
Used in extremely precise work when delicate hand operation is required
115mm
33mm
7mm