Learn More
DWK Life Sciences Wheaton™ Natural Polypropylene Caps for Narrow-Mouth Containers Available on GSA/VA Contract for Federal Government customers only.
For use with Wheaton leak-resistant narrow mouth bottles
Manufacturer:  DWK Life Sciences 239506
 View more versions of this product
Catalog No. 09-877-11A
Add to cart
Description
Description
  • Leak-proof design
  • Autoclavable
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
ISO 9001
Polypropylene
Redi-pak™ leak resistant narrow mouth bottle
Natural
72 Cs.
20-410