Wheaton™ Plastic Nongraduated Media Bottles

Sterile and versatile

Manufacturer: Wheaton™ 219979

 View more versions of this product

Catalog No. 06-451-299

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Specifications

    Sterile Yes
    Certifications/Compliance USP Type 1
    Closure Material Polypropylene
    Color Clear
    Diameter (English) 2.95 in.
    Graduated No
    Height (Metric) 175mm
    Material PET
    Shape Square
    Capacity (Metric) 500mL
    Closure Size 38-430
    Diameter (Metric) 75mm
    Height (English) 6.88 in.
    Applications For distilled Water, analytical standards and reagents when fitted with a Polyethylene lined screw cap
    Neck Finish 33-430
    Quantity 12 Cs.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit