Wheaton™ Polypropylene Caps for Lab 45 Graduated Bottles

Replacement caps for Lab 45 media bottles

Manufacturer: Wheaton™ W24072605

 View more versions of this product

Catalog No. 02-926-980

  Check Availability
Add to Cart

    Specifications

    Certifications/Compliance ISO 9001
    Color Pink
    For Use With Lab 45 graduated bottle
    Includes Wedge shaped inner sealing ring
    Material Polypropylene
    No. per Case 12 Cs.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit