Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Wheaton™ Polypropylene Caps for Lab 45 Graduated Bottles
Click to view available options
Color:
Blue
Red
White
Includes:
Cap without inner sealing ring
PTFE/Silicone Liner
Wedge shaped inner sealing ring
Description
- 45mm screw thread
- Polypropylene
- Autoclavable
- Linerless
Specifications
For Use With (Equipment) | Lab 45 graduated bottle |
Color | White |
Includes | Cap without inner sealing ring |
Material | Polypropylene |
No. per Case | 12 Cs. |
Certifications/Compliance | ISO 9001 |
Autoclavable | Autoclavable |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction