Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Polypropylene Caps for Wheaton Dropping Bottles - Blue

Click to view available options
Neck Finish:
13-425
15-415
20-410
8-425
No. per Case:
100/Cs.
1000/Cs.
  • Packaged in case of 100 or 1000
For Use With (Equipment) Plastic dropping bottle
Certifications/Compliance ISO 9001
Color Blue
Material Polypropylene
No. per Case 1000/Cs.
Type Dropping Bottle
Neck Finish 13-425