Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Polypropylene Caps for Wheaton Dropping Bottles - Pink
SDP

Catalog Number 0344013
Click to view available options
Neck Finish:
13-425
15-415
20-410
8-425
For Use With (Equipment):
Dropping bottle
Plastic dropping bottle
No. per Case:
100 Cs.
1000 Cs.

Screw cap made of radiation grade polypropylene

  • Packaged in case of 100 or 1000
For Use With (Equipment) Plastic dropping bottle
Product Type Polypropylene Cap
Color Pink
Material Polypropylene
No. per Case 1000 Cs.
Neck Finish 15-415
Certifications/Compliance ISO 9001
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit