Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Wheaton™ Polypropylene Caps for Wheaton Dropping Bottles - Yellow
Click to view available options
Neck Finish:
13-425
15-415
20-410
8-425
For Use With (Equipment):
Dropping bottle
Plastic dropping bottle
No. per Case:
100 Cs.
1000 Cs.
Description
- Packaged in case of 100 or 1000
Specifications
For Use With (Equipment) | Plastic dropping bottle |
Product Type | Polypropylene Cap |
Color | Yellow |
Material | Polypropylene |
No. per Case | 100 Cs. |
Neck Finish | 13-425 |
Certifications/Compliance | ISO 9001 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction