Learn More
DWK Life Sciences Wheaton™ Prescored Clear Standards Ampules Available on GSA/VA Contract for Federal Government customers only.
Glass containers designed to be hermetically sealed, preserving purity and extending content shelf life
Manufacturer:  DWK Life Sciences 176759
 View more versions of this product
Catalog No. 01-215E
Add to cart
Description
Description
A gold band on the stem indicates that the ampule is pre-scored to facilitate snapping.
  • Constructed of USP Type I clear and amber low-extractable borosilicate glass
  • Methods of sterilization: autoclave, dry heat and EtO
  • Prevents cross contamination
Standards packaging, environmental and diagnostic standards, lyophilization and cryogenic storage
This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.
Specifications
Specifications
7mL
Clear
1.7cm
9.5cm
Prescored Standard Ampule
ASTM E438 Type I, Class A
0.66 in.
3.74 in.
Borosilicate Glass