Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ R2P™ Roller Culture Apparatus

Go from research to production without having to change protocols and revalidate

$1121.66

Specifications

Certifications CE, CSA, UL
Controller Type R2P
Hertz 50/60Hz
Voltage 120V
Wattage 35w
View More Specs

Products 1
Catalog Number Mfr. No. Capacity (English) Speed Range Height (Metric) Exterior Price Quantity & Availability  
Catalog Number Mfr. No. Capacity (English) Speed Range Height (Metric) Exterior Price Quantity & Availability  
02-911-550
View Documents
DWK Life Sciences
W348888
5 Bottle Positions 0.25 to 8.0rpm 15.2cm
Case for $1,121.66
Add to cart
 
Description

Description

The 88-Position R2P Large-Scale Production Apparatus is ideal for pharmaceutical or vaccine production. The R2P production and modular systems allow you to go from research to production without having to change protocols and revalidate; simply add decks and optional features as needed.

Key Features

  • Large digital display and keypad enable easy input and monitoring of settings
  • Units continually display the bottle rpm setting and actual bottle rpm
  • Programmable soft start and stop feature avoids premature cell detachment; settings can be saved in one of ten memory locations for easy recall
  • Rotational speed and drivetrain are continuously monitored; built-in rotation alarm can be tied to an existing monitoring system and provides both visual and audible warning of out-of-tolerance conditions All-position drive accepts glass or plastic bottles 110 to 121mm in diameter and up to 550mm long
  • Optional CART2™ computer software monitors, controls, and records data on up to 2047 units

Holds up to 100 bottles

  • With only 6 in. (15.2cm) between decks, it can accommodate up to 110 850 cm2 bottles on eleven decks and still fit into most standard incubators, including The Breeder™ (22-288-530), and Wheaton Roll-In Incubator (11-690-1A)
  • Weight of drive unit with one deck: 53 lb. (24kg); each additional deck is 161/2 lb. (7.5kg)
  • Dimensions of drive unit with one deck is 241/2L x 31W x 131/2 in.H (62 x 79 x 34.5cm); each additional deck is 6 in.H (15.2cm)
Battery Backup Option
  • Monitors the AC power and automatically switches between battery power and AC power as needed
  • Provides auxiliary power to maintain complete functionality of the R2P unit for a minimum of 18 hours
  • Can be used to rotate the roller vessels during transport to and from the warm room or incubator
  • Consists of two long-life sealed batteries and an automatic charger system
  • Audible alarm and display backlight will not function during battery power mode
  • Based on 110mm diameter vessels rolling at 5rpm
  • Recharge time is 12 hours for completely discharged batteries, shorter for partially discharged batteries
  • Automatically monitors temperature and adjusts charge voltage for correct charging levels at elevated incubator temperatures

Additional configurations are available; contact your Fisher Scientific Customer Service Representative for more information

Order Info

Electrical requirements: 110/120VAC, 50/60Hz, 35w

Certifications

Conform to UL, CSA, and CE Standards

Specifications

Specifications

CE, CSA, UL
50/60Hz
35w
R2P
120V
120V 50/60Hz
SDS
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit