Wheaton™ Replacement O-Rings for Dry-Seal Vacuum Desiccator Available on GSA/VA Contract for Federal Government customers only.
Manufacturer: Wheaton™ 366024
 View more versions of this product
Catalog No. 08-634-4B
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    For Use With (Equipment) For dry-seal vacuum desiccator
    Diameter (English) Outer 6 in.
    For Use With (Application) Createing an airtight seal when vacuum is applied to dessicator
    Diameter (Metric) Outer 150mm
    Material Silicone Rubber
    Product Type Replacement O-Ring
  • Description & Specifications
Related Products
Product Content Feedback