Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Wheaton™ Round Translucent LDPE Bottles
Click to view available options
Capacity:
0.24 oz., 7 mL
0.5 oz., 15 mL
1 oz., 30mL
Height (English):
2.4 in.
2.87 in.
3.07 in.
Height (Metric):
63 mm
73 mm
78 mm
Neck Finish:
15-415
20-410
Description
Narrow mouth is ideal for liquids.
- Excellent for mild and strong buffers, good chemical resistance
- Pre-attached white polypropylene screw cap with foamed polyethylene liner
Specifications
Closure Type | Screw Cap |
Closure Liner | Polyethylene Foam |
Closure Material | Polyethylene |
Closure Size | 15-415 mm |
Diameter (Metric) | 19 mm |
Height (English) | 2.4 in. |
Height (Metric) | 63 mm |
Capacity | 0.24 oz., 7 mL |
Capacity (Metric) | 7 mL |
Capacity (English) | 0.24 oz. |
Show More |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction