Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wheaton™ CryoELITE™ Cryogenic Storage Vials

CryoELITE Vial with Loctagon skirt provides stability in freestanding position, and locks into the CryoELITE Benchmate Rack, making it easy to open and close the vial with one hand.

Manufacturer:  Wheaton™ W985866

 View more versions of this product

Catalog No. 02-912-731


  Check Availability
Add to cart

Specifications Description & Specifications

Specifications

Yellow
Yellow
2mL
Polypropylene
External
1.92 in.
Marking Spot
Yes
DOT, IATA
0.47 in.
12mm
Yes
Yes
49mm
500/Case
Cryogenic Vial
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit