Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Wheaton™ Large Safety-Coated Solution and Serum Bottles

Click to view available options
Capacity (Metric):
18927 mL

Plastisol coated for safety.

  • Material: Borosilicate Glass
  • Not autoclavable
  • Shape: Round
  • Takes rubber stopper No. 8
  • Light resistant
Height (Metric) 502 mm
Closure Material Rubber
Diameter (Metric) 294 mm
Height (English) 19.76 in.
Capacity 640 oz., 18927 mL
Capacity (Metric) 18927 mL
Capacity (English) 640 oz.
Color Clear
Diameter (English) 11.57 in.
Material Borosilicate Glass
Type Serum Bottle
Show More Show Less