Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Wheaton™ White Polypropylene Caps With PTFE Faced Foamed Polyethylene Liner
Click to view available options
Neck Finish:
13-425
15-425
Description
- Can withstand autoclaving / sterilization
Specifications
Closure Liner | PTFE/Polyethylene |
Product Type | Polypropylene Cap |
For Use With (Equipment) | Glass bottle or vial |
Color | White |
Material | Polypropylene |
No. per Case | 144 Cs. |
Neck Finish | 13-425 |
Certifications/Compliance | ISO 9001 |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction