Learn More
DWK Life Sciences Wheaton™ Screw Thread Headspace Closures
Manufacturer:  DWK Life Sciences W224220
 View more versions of this product
Catalog No. 03-410-734
  Check Availability
Add to cart
Description
Description
  • For use with CTC Analytics Systems requiring magnetic closures
  • Also compatible with modern Agilent, Perkin Elmer and Tekmar autosamplers that do not require transports
Specifications
Specifications
Red PTFE/White Silicone
Screw Cap