Wheaton™ Self-Refilling Repetitive Syringes, Microtest Manifolds
Manufacturer: Wheaton™ 851380
 View more versions of this product
Catalog No. 13-708-1
  Check Availability
Add to Cart
    Specifications
    Accuracy ±6%
    Angle Straight
    Autoclavable Yes
    For Use With (Equipment) Self refilling repetitive syringes, 24 and 96-well plates, Acura™ 865 microdispenser pipette
    Material Stainless Steel
    Number of Channels 4.0
    Product Type Microtest manifold with PTFE stopper
    No. of Positions 4.0
    Spacing 20mm
  • Description & Specifications
Related Products
Product Content Feedback