Wheaton™ Serum Tubing Vials Available on GSA/VA Contract for Federal Government customers only.

Available as clear and amber vials

Manufacturer: Wheaton™ 223684

 View more versions of this product

Catalog No. 06-446C

  Check Availability
Add to Cart

    Specifications

    Autoclavable Yes
    Diameter (English) Inside 0.27 in.
    Diameter (Metric) Inside 7mm
    Diameter (Metric) Outside 17mm
    Color Clear
    Certifications/Compliance USP Type I, ASTM E438, Type I, Class A
    Material Borosilicate Glass
    Type Serum Tubing Vial
    Volume (Metric) 3mL
    Closure Type Crimp
    Height (Metric) 38mm
    Capacity (Metric) 3.0mL
    For Use With (Application) For lyophilization, packaging, sampling and freeze drying applications
    Volume (English) 0.8 dr.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit