Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Wheaton™ Serum Vials
Available as clear and amber vials
$253.50
Specifications
Material | Borosilicate Glass |
---|---|
For Use With (Application) | For lyophilization, packaging, sampling and freeze drying applications |
Autoclavable | Autoclavable |
Closure Type | Crimp Cap |
Certifications/Compliance | ASTM E438, USP Type I, Type I, Class A |
Catalog Number | Mfr. No. | Color | Diameter (Metric) Outer | Volume (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Color | Diameter (Metric) Outer | Volume (Metric) | Price | Quantity & Availability | |||
06-446E
|
DWK Life Sciences
223687 |
Clear | 30.5 mm | 20 mL |
Case of 120 for $253.50
|
|
|||
06-406-37
|
DWK Life Sciences
223683 |
Clear | 15 mm | 2 mL |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | |||
06-446C
|
DWK Life Sciences
223684 |
Clear | 17 mm | 3 mL |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | |||
06-446D
|
DWK Life Sciences
223685 |
Clear | 22 mm | 5 mL |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | |||
06-446G
|
DWK Life Sciences
223693 |
Amber | 15 mm | 2 mL |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | |||
06-446H
|
DWK Life Sciences
223695 |
Amber | 22 mm | 5 mL |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | |||
06-446J
|
DWK Life Sciences
223696 |
Amber | 24 mm | 10 mL |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | |||
06-406-38
|
DWK Life Sciences
223686 |
Clear | 24 mm | 10.0 mL |
Item Discontinued
This item has been discontinued by the supplier. Please Sign In to view product availability in your area. View alternate product |
N/A | |||
Description
Available as clear and amber vials
Glass can withstand the challenges of reduced pressure and extreme temperature variations. Protects against changes in pH and maintain the purity of the contents.
- Clear vials made from low extractable borosilicate glass tubing to ensure uniform wall thickness
- Amber vials made from borosilicate glass conforms to USP Type 1 requirements to protect light-sensitive samples
- Withstand extreme temperature variations
- Necks finished for aluminum crimp seals
- Blowback feature of neck and specially designed bottom radius for lyophilization applications
- Packaged in shrink-wrapped modules to reduce particulate contamination
Lyophilization, packaging, and sampling
Compliance
Low extractable borosilicate glass meets ASTM™ Type 1, Class A and USP Type 1 standards
Specifications
Borosilicate Glass | |
Autoclavable | |
ASTM E438, USP Type I, Type I, Class A |
For lyophilization, packaging, sampling and freeze drying applications | |
Crimp Cap |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title