Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences™ Wheaton™ Serum Vial
Shop All DWK Life Sciences Products

Click to view available options
Volume (Metric):
2 mL
3 mL
4 mL
5 mL
10 mL
13.5 mL
Color:
Amber
Clear
Quantity:
10 Vials/Pk., 100 Pk./Cs.
100 Vials/Pk.
100 Vials/Pk., 10 Pk./Cs.
100 Vials/Pk., 304 Pk./Cs.
12 Vials/Pk., 196 Pk./Cs.
150 Vials/Pk.
150 Vials/Pk., 14 Pk./Cs.
150 Vials/Pk., 264 Pk./Cs.
16 Vials/Pk., 150 Pk./Cs.
169 Vials/Pk.
169 Vials/Pk., 10 Pk./Cs.
169 Vials/Pk., 304 Pk./Cs.
225 Vials/Pk.
225 Vials/Pk., 12 Pk./Cs.
225 Vials/Pk., 304 Pk./Cs.
288 Vials/Pk., 150 Pk./Cs.
304 Vials/Pk., 100 Pk./Cs.
304 Vials/Pk., 196 Pk./Cs.
Description
- Vials conform to the hydrolytic resistance requirements outlined in USP 660 and EP 3.2.1
- Shrink-wrapped modules reduce particulate contamination
- Special design adds strength for freeze-drying applications
Specifications
Specifications
| Color | Clear |
| Diameter (Metric) Outer | 20 mm |
| Material | Borosilicate Glass |
| Pre-slit | No |
| Sterility | Non-sterile |
| Volume (Metric) | 5 mL |
| Autoclavable | Autoclavable |
| Height (Metric) | 40 mm |
| Quantity | 100 Vials/Pk., 304 Pk./Cs. |
| Type | Serum Vial |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction